Merck
  • Identification of neighboring protein pairs in the 60 S ribosomal subunits from Saccharomyces cerevisiae by chemical cross-linking.
Identification of neighboring protein pairs in the 60 S ribosomal subunits from Saccharomyces cerevisiae by chemical cross-linking.
The Journal of biological chemistry (1989-06-25)
R H Xiang, J C Lee
ABSTRACT
Protein-protein cross-linking was used to determine the spatial arrangement of proteins within the 60 S ribosomal subunits of Saccharomyces cerevisiae. Protein cross-links were generated by treatment of intact ribosomal subunits with dimethyl 3,3'-dithiobispropionimidate. Proteins were extracted from the treated subunits and fractionated by Cm-cellulose chromatography. Cross-linked proteins in these fractions were analyzed by electrophoresis on two-dimensional diagonal polyacrylamide gels containing sodium dodecyl sulfate. Component members of cross-linked pairs were radiolabeled with 125I and identified by two-dimensional gel electrophoresis and comparison with nonradioactive ribosomal protein markers. Seventeen pairs involving 16 of the 45 60 S subunit proteins were identified. Several proteins were detected in numerous cross-linked dimers and were used as foci for constructing a model depicting the arrangement of proteins within the 60 S ribosomal subunit. The model also incorporated previously published data on structure and function of proteins from the yeast 60 S subunit.
Sigma-Aldrich
Dimethyl 3,3′-dithiopropionimidate dihydrochloride, powder