Merck
  • High-performance liquid chromatography separation of phospholipid classes and arachidonic acid on cyanopropyl columns.
High-performance liquid chromatography separation of phospholipid classes and arachidonic acid on cyanopropyl columns.
Analytical biochemistry (1989-10-01)
J M Samet, M Friedman, D C Henke
ABSTRACT
An HPLC method for the separation and analysis of arachidonic acid and eight phospholipid classes is described: phosphatidylglycerol, phosphatidylinositol, cardiolipin, phosphatidylserine, phosphatidylethanolamine, phosphatidylcholine, sphingomyelin, and 2-lysophosphatidylcholine. The separation is carried out at 60 degrees C on 2 cyanopropyl columns using a gradient of acetonitrile and 5 mM sodium acetate (pH 5.0). Cyanopropyl columns require a lower proportion of water in the mobile phase to elute the more polar phospholipids than other types of columns and are thus less prone to equilibration problems. The method is highly reproducible (average coefficient of variation for each retention time less than or equal to 3.5%) and permits analysis of peaks by phosphorus content. Data obtained by analyzing lipid extracts from rat alveolar macrophages prelabeled with [G-3H]-arachidonic acid were analyzed by this HPLC method and compared to standard analysis by TLC. There was a significant correlation between the radioactivity profiles obtained with the two chromatographic methods (HPLC versus TLC) by linear regression analysis [HPLC = 0.83 (TLC) + 3.58, n = 25, r = 0.95, P less than 0.001].
Sigma-Aldrich
Butyronitrile, purum, ≥99.0% (GC)
Sigma-Aldrich
Butyronitrile, ≥99%