Merck
  • Imidazole-induced contractility of vascular smooth muscle cells in the presence of U-73122, ODQ, indomethacin and 7-nitroindazole.
Imidazole-induced contractility of vascular smooth muscle cells in the presence of U-73122, ODQ, indomethacin and 7-nitroindazole.
Polish journal of veterinary sciences (2013-08-27)
M Wiciński, G Grześk, B Malinowski, E Grześk, A T Sinjab, M Krzyzanowski, A Michalska, D Bylina, J Dziecioł, K Szadujkis-Szadurska, A Czeczuk
ABSTRACT
The aim of the study was to assess the impact of modulating factors on vascular smooth muscle cells reactivity. Vascular resistance was induced by the administration of increasing concentrations of imidazole. The experiments were performed on isolated and perfused tail artery of Wistar rats (weight 250 g - 350 g). Rats were been narcotized by urethane (intraperitoneal injection) at a dose of 120 mg/kg, stunned and then sacrificed by cervical dislocation. In the following investigation classical pharmacometric methods were used. Relationships between concentration-response curves (CRCs) for imidazole observed in the presence of ODQ [(1H-(1,2,4)oxadiazolo-[4,3-a]quinoxalin-1-one)], 7-nitroindazole and indomethacin were analyzed. Imidazole-induced contractility of vascular smooth muscle cells was independent from alpha-adrenergic receptors and PLC activity. Reactivity of VSMCsinduced by imidazole, was significantly changed in the presence of ODQ and 7-nitroindazole.
Sigma-Aldrich
Imidazole, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Imidazole, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Imidazole, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
Imidazole, ReagentPlus®, 99%
Sigma-Aldrich
7-Nitroindazole, ≥98%
Sigma-Aldrich
Indomethacin, meets USP testing specifications
Sigma-Aldrich
Imidazole hydrochloride
Sigma-Aldrich
Imidazole, for molecular biology, ≥99% (titration)
Sigma-Aldrich
Imidazole, ≥99% (titration), crystalline
Sigma-Aldrich
Imidazole, ACS reagent, ≥99% (titration)
Sigma-Aldrich
Indomethacin, 98.5-100.5% (in accordance with EP)
Sigma-Aldrich
Imidazole sodium derivative, technical grade
Imidazole, European Pharmacopoeia (EP) Reference Standard
Indomethacin, European Pharmacopoeia (EP) Reference Standard
Supelco
Indomethacin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Imidazole, ReagentPlus®, 99%, Redi-Dri, free-flowing
Supelco
Imidazole, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Imidazole buffer Solution, BioUltra, 1 M in H2O