Merck
  • Association of NALP2 polymorphism with arsenic induced skin lesions and other health effects.
Association of NALP2 polymorphism with arsenic induced skin lesions and other health effects.
Mutation research (2013-05-07)
Pritha Bhattacharjee, Nandana Das, Debmita Chatterjee, Anirban Banerjee, Jayanta K Das, Santanu Basu, Saptarshi Banerjee, Papia Majumder, Prashant Goswami, Ashok K Giri
ABSTRACT
Prolonged consumption of arsenic-laden water above the threshold limit of 10μg/L causes a plethora of dermatological and non-dermatological multi-organ health problems, including cancer and death. Among several mechanisms of arsenic-induced toxicity and carcinogenicity studied so far, role of arsenic in impairment of immune system is less understood. Epidemiological data, animal model as well as cell line based studies have indicated that arsenic targets immune system and is associated with characteristic immunosupression, which may further adversely affect respiratory function. However, to the best of our knowledge, there is no study with respect to arsenic susceptibility investigating the role of genetic variation having immunological function. Hence, we have recruited a total of 432 arsenic-exposed individuals, of which 219 individuals with characteristic arsenic-induced skin lesions (cases) and 213 individuals without arsenic-induced skin lesion(controls), from arsenic-exposed districts of West Bengal, India. To find any probable association between arsenicism and the exonic single nucleotide polymorphisms (SNPs) in NALP2 gene, an important component of inflammasome complex, we screened the entire coding region (exon) in all the study participants. Among 9 SNPs found in NALP2 gene, the A1052E polymorphism (at least with one minor allele), was significantly overrepresented in controls and hence implies decreased risk toward the development of skin lesions [OR=0.67, 95% CI: 0.46-0.97]. Since, development of non-dermatological health effects are also important factor to properly look into, we have attempted to correlate the genetic variation of NALP2 with the extent of cytogenetic damage as measured by chromosomal aberration assay and adverse health effects including peripheral neuropathy, eye problem and respiratory diseases in the study population. We observed individuals with the protective genotype had less chromosomal aberration (p<0.05), and were also less susceptible toward arsenic-related respiratory diseases [OR=0.47; 95%CI: 0.23-0.89]. These findings suggest that NALP2 A1052E SNP plays an important role toward development of arsenic-induced skin lesions, chromosomal damage and respiratory diseases.
Sigma-Aldrich
Arsenic, 99.999% trace metals basis
Sigma-Aldrich
Arsenic, powder, ≥99.997% trace metals basis