Merck
  • Potent antitrypanosomal activities of heat shock protein 90 inhibitors in vitro and in vivo.
Potent antitrypanosomal activities of heat shock protein 90 inhibitors in vitro and in vivo.
The Journal of infectious diseases (2013-05-01)
Kirsten J Meyer, Theresa A Shapiro
ABSTRACT
African sleeping sickness, caused by the protozoan parasite Trypanosoma brucei, is universally fatal if untreated, and current drugs are limited by severe toxicities and difficult administration. New antitrypanosomals are greatly needed. Heat shock protein 90 (Hsp90) is a conserved and ubiquitously expressed molecular chaperone essential for stress responses and cellular signaling. We investigated Hsp90 inhibitors for their antitrypanosomal activity. Geldanamycin and radicicol had nanomolar potency in vitro against bloodstream-form T. brucei; novobiocin had micromolar activity. In structure-activity studies of geldanamycin analogs, 17-AAG and 17-DMAG were most selective against T. brucei as compared to mammalian cells. 17-AAG treatment sensitized trypanosomes to heat shock and caused severe morphological abnormalities and cell cycle disruption. Both oral and parenteral 17-DMAG cured mice of a normally lethal infection of T. brucei. These promising results support the use of inhibitors to study Hsp90 function in trypanosomes and to expand current clinical development of Hsp90 inhibitors to include T. brucei.
Product Description
Supelco
Novobiocin sodium salt, VETRANAL®, analytical standard
Sigma-Aldrich
Novobiocin sodium, meets USP testing specifications
Sigma-Aldrich
Novobiocin sodium salt, ≥90% (HPLC)
Sigma-Aldrich
Radicicol from Diheterospora chlamydosporia, solid