Merck
  • Successful treatment of severe amiodarone pulmonary toxicity with polymyxin B-immobilized fiber column direct hemoperfusion.
Successful treatment of severe amiodarone pulmonary toxicity with polymyxin B-immobilized fiber column direct hemoperfusion.
Chest (2013-04-03)
Nahoko Sato, Keisuke Kojima, Yuko Horio, Eisuke Goto, Aiko Masunaga, Hidenori Ichiyasu, Hirotsugu Kohrogi
ABSTRACT
Amiodarone pulmonary toxicity (APT) is the most serious side effect of amiodarone. Although severe APT, such as ARDS, is rare, mortality of severe APT is high. Polymyxin B-immobilized fiber column direct hemoperfusion (PMX-DHP) is a medical device that reduces blood endotoxin levels in sepsis. Recent reports have shown that PMX-DHP improves oxygenation in patients with acute exacerbation of idiopathic pulmonary fibrosis and drug-induced severe interstitial pneumonia. Here, we present a case study of a patient with severe APT treated with PMX-DHP with complete recovery. The patient rapidly developed respiratory failure and required mechanical ventilation. Despite corticosteroid pulse therapy, no clinical improvement was noted. PMX-DHP was then started, and severe respiratory failure improved with reduction of serum levels of amiodarone and its metabolite monodesethylamiodarone. The patient was weaned from mechanical ventilation and has done well without recurrence. To our knowledge, this is the first reported case of PMX-DHP therapy for severe APT. We speculate that PMX-DHP could be a new treatment strategy for severe APT.
Sigma-Aldrich
Polymyxin B sulfate, meets USP testing specifications
Polymyxin B sulfate, European Pharmacopoeia (EP) Reference Standard
Supelco
Polymyxin B solution, 1 mg/mL in H2O, analytical standard
Sigma-Aldrich
Polymyxin B sulfate salt, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Polymyxin B sulfate salt
Sigma-Aldrich
Polymyxin B solution, 20 mg/mL in H2O