Merck
  • Differential toxicity biomarkers for irinotecan- and oxaliplatin-containing chemotherapy in colorectal cancer.
Differential toxicity biomarkers for irinotecan- and oxaliplatin-containing chemotherapy in colorectal cancer.
Cancer chemotherapy and pharmacology (2013-04-02)
Lucía Cortejoso, María I García, Pilar García-Alfonso, Eva González-Haba, Fernando Escolar, María Sanjurjo, Luis A López-Fernández
ABSTRACT
Oxaliplatin or irinotecan is usually administered jointly with fluoropyrimidines in colorectal cancer patients treated with chemotherapy. Both drugs have different toxicity patterns. Biomarkers for predicting high-risk severe adverse reactions can help select the best treatment. A retrospective analysis of 106 colorectal cancer patients receiving an oxaliplatin-based treatment and 56 receiving an irinotecan-based treatment was performed. One copy number variant (GSTT1) and nine polymorphisms in irinotecan and oxaliplatin metabolism, transport or DNA repair genes (ABCB1, UGT1A1, XRCC1, ERCC1, ERCC2, GSTP1) were genotyped by SNaPshot, polymerase chain reactions' length fragments, or copy number assays. In irinotecan-treated patients, T allele of ABCB1C1236T SNP was associated with a lower risk of asthenia(OR = 0.047; 95 % CI = 0.004–0.493; P = 0.011) and Tallele of ABCB1 C3435T SNP was associated with a lower risk of diarrhea (OR = 0.177; 95 % CI = 0.034–0.919;P = 0.039), and individuals with two copies of GSTT1 gene had a lower risk for asthenia (OR = 0.093; 95 %CI = 0.011–0.794; P = 0.030). In oxaliplatin-treated patients, carriers of one or two T variants of Asn118Asn ERCC1 SNP had a lower risk for neutropenia(OR = 0.205; 95 % CI = 0.061–0.690; P = 0.01) [corrected]. These biomarkers could help oncologists select the best treatment by reducing toxicity associated with irinotecan or oxaliplatin in colorectal cancer patients, thus increasing their quality of life.
Sigma-Aldrich
Irinotecan hydrochloride, topoisomerase inhibitor
Oxaliplatin, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
7-Ethyl-10-hydroxycamptothecin, ≥98% (HPLC), powder
Sigma-Aldrich
Oxaliplatin, powder