Merck
  • The long-term effect of nitrite on the granule-based enhanced biological phosphorus removal system and the reversibility.
The long-term effect of nitrite on the granule-based enhanced biological phosphorus removal system and the reversibility.
Bioresource technology (2013-02-23)
Xiongliu Zheng, Peide Sun, Juqing Lou, Zhiguo Fang, Maoxin Guo, Yingqi Song, Xiudi Tang, Tao Jiang
ABSTRACT
This study investigated the long-term effect of nitrite on the granule-based enhanced biological phosphorus removal (EBPR) system and the reversibility from macro- to micro-scale. Nitrite was found to seriously deteriorate the EBPR performance and result in severe sludge bulking. The inhibited polysaccharides excretion could lead to breaking the stability and integrity of the granules. Therefore, the reduced particle size and granule disintegration were observed. In this study, granules with lower ratio of proteins to polysaccharides (1.76) had better structure and function than the higher (3.84). Experimental results demonstrated that the microbial community structure was largely changed due to the presence of nitrite. In comparison, glycogen accumulating organisms (GAOs) had stronger resistibility and higher recovery rate than poly-phosphate accumulating organisms (PAOs). Interestingly, the community composition was unable to recover (Dice coefficients, 33.0%), although good EBPR performance was achieved only by propagating other types of PAOs.
Sigma-Aldrich
Glycogen from bovine liver, ≥85% dry basis (enzymatic)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Glycogen from oyster
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Glycogen from mussel, Mytilus genus, ≥85% anhydrous basis (enzymatic)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Glycogen from mussel, Mytilus genus, for DNA precipitations
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Glycogen from rabbit liver, ≥85% dry basis (enzymatic)