- Delphinidin inhibits IL-1β-induced activation of NF-κB by modulating the phosphorylation of IRAK-1(Ser376) in human articular chondrocytes.
Delphinidin inhibits IL-1β-induced activation of NF-κB by modulating the phosphorylation of IRAK-1(Ser376) in human articular chondrocytes.
In OA, there is enhanced expression of pro-inflammatory cytokines such as IL-1β in the affected joint. Delphinidin, an anthocyanidin found in pigmented fruits and vegetables, has been shown to possess anti-inflammatory and antioxidant properties. In the present study we determined whether delphinidin would inhibit the IL-1β-induced activation of NF-κB in human chondrocytes and determined the mechanism of its action. PGE2 levels and activation of NF-κB p65 in human OA chondrocytes were determined by ELISA-based assays. Protein expression of cyclo-oxygenase-2 (COX-2) and phosphorylation of kinases was determined by western immunoblotting. Expression level of mRNAs was determined by TaqMan assays. Delphinidin inhibited IL-1β-induced expression of COX-2 and production of PGE2 in human chondrocytes. Delphinidin also inhibited IL-1β-mediated phosphorylation of IL-1 receptor-associated kinase-1(Ser376), phosphorylation of IKKα/β, expression of IKKβ, degradation of IκBα, and activation and nuclear translocation of NF-κB/p65. Phosphorylation of TGF-β-activated kinase 1 was not observed but NF-κB-inducing kinase (NIK) was phosphorylated and phosphorylation of NIK was blocked by delphinidin in IL-1β-treated human chondrocytes. These data identify delphinidin as a novel inhibitor of IL-1β-induced production of cartilage-degrading molecule PGE2 via inhibition of COX-2 expression and provide new insight into the mechanism of its action. Our results also identify inhibition of IRAK1(Ser376) phosphorylation by delphinidin in IL-1β-induced activation of NF-κB in human chondrocytes. Given the important role played by IL-1β-induced NF-κB activation, COX-2 expression and PGE2 production in OA, our results may have important implications for the development of novel therapeutic strategies for the prevention/treatment of OA.