Merck
  • Laser-assisted growth of t-Te nanotubes and their controlled photo-induced unzipping to ultrathin core-Te/sheath-TeO(2) nanowires.
Laser-assisted growth of t-Te nanotubes and their controlled photo-induced unzipping to ultrathin core-Te/sheath-TeO(2) nanowires.
Scientific reports (2013-02-06)
Thomas Vasileiadis, Vassileios Dracopoulos, Mary Kollia, Spyros N Yannopoulos
ABSTRACT
One dimensional (1D) nanostructures of semiconducting oxides and elemental chalcogens culminate over the last decade in nanotechnology owing to their unique properties exploitable in several applications sectors. Whereas several synthetic strategies have been established for rational design of 1D materials using solution chemistry and high temperature evaporation methods, much less attention has been given to the laser-assisted growth of hybrid nanostructures. Here, we present a laser-assisted method for the controlled fabrication of Te nanotubes. A series of light-driven phase transition is employed to controllably transform Te nanotubes to core-Te/sheath-TeO(2) and/or even neat TeO(2) nanowires. This solid-state laser-processing of semiconducting materials apart from offering new opportunities for the fast and spatially controlled fabrication of anisotropic nanostructures, provides a means of simultaneous growing and integrating these nanostructures into an optoelectronic or photonic device.
Sigma-Aldrich
Tellurium dioxide, ≥97.0%
Sigma-Aldrich
Tellurium, granular, −5-+50 mesh, 99.99% trace metals basis
Sigma-Aldrich
Tellurium, powder, −200 mesh, 99.8% trace metals basis
Sigma-Aldrich
Tellurium dioxide, ≥99%
Sigma-Aldrich
Tellurium dioxide, 99.995% trace metals basis
Sigma-Aldrich
Tellurium, powder, −30 mesh, 99.997% trace metals basis
Sigma-Aldrich
Tellurium, pieces, 99.999% trace metals basis
Sigma-Aldrich
Tellurium, shot, ≤2 mm, 99.999% trace metals basis