Merck
  • Repeat dose opioids may be effective for breathlessness in chronic heart failure if given for long enough.
Repeat dose opioids may be effective for breathlessness in chronic heart failure if given for long enough.
Journal of palliative medicine (2013-02-02)
Stephen G Oxberry, J Martin Bland, Andrew L Clark, John G Cleland, Miriam J Johnson
ABSTRACT
The longer-term effects of opioids for breathlessness are not known in people with chronic heart failure (CHF). Our aim was to assess the longer-term effect of oral opioids on breathlessness due to CHF. We conducted a 3-month open-label extension to a crossover randomized controlled trial (RCT) comparing 4 days of morphine, oxycodone, and placebo. Thirty-five participants from a tertiary cardiology clinic completed the RCT. Thirty-three were followed for 3 months, continuing open-label opioids if they wished. Thirteen participants continued an opioid; 20 did not. Four measures of breathlessness intensity (0-10 numerical rating scale [NRS] and modified Borg score, each recording worst and average breathlessness during 24 hours) were combined using principal component analysis to give a single measure for the primary analysis. Groups were compared using analysis of covariance. Secondary measures included quality of life (SF-12(®) Health Survey), cardiorespiratory, and global impression of change in breathlessness at 3 months. At 3 months, the composite breathlessness measure improved to a greater extent in the opioid group (p=0.017). The opioid group had an improvement in global impression of change (mean 2.62 [opioids] versus -0.65 [nonopioids]; p=0.0009). The SF-12 physical component improved more in the opioid group (p=0.014). Cardiorespiratory variables were unchanged. Opioids given for 3 months were well tolerated and safe. Opioid-related improvement in breathlessness in people with CHF might not be seen until longer-term administration. We cannot conclude from these data that they are effective and a longer-term RCT is needed.
Product Description
Supelco
Oxycodone solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
Oxycodone hydrochloride, analytical standard