Merck
  • Tuning the electrocatalytic hydrogen evolution reaction promoted by [Mo2O2S2]-based molybdenum cycles in aqueous medium.
Tuning the electrocatalytic hydrogen evolution reaction promoted by [Mo2O2S2]-based molybdenum cycles in aqueous medium.
Dalton transactions (Cambridge, England : 2003) (2013-02-01)
Akram Hijazi, Justin Claude Kemmegne-Mbouguen, Sébastien Floquet, Jérôme Marrot, Jennifer Fize, Vincent Artero, Olivier David, Emmanuel Magnier, Bruce Pégot, Emmanuel Cadot
ABSTRACT
We report the syntheses and characterizations, in the solid state and in solution, of three new cyclic polyoxothiomolybdates self-assembled around 2,5-dimethylterephthalate (DMT) and 2,5-bis(trifluoromethyl)terephthalate (DFMT) ligands, namely [Mo12DMT](2-), [Mo12DFMT](2-) and [Mo16DFMT](2-). A series of these two Mo12-compounds completed by the two Mo12-compounds obtained with 2,3,5,6-tetramethylterephthalate and trimesate ligands offer the opportunity to compare their electro-catalytic properties for reduction of protons into hydrogen. The ability of these compounds to promote the reduction of protons into hydrogen in aqueous medium is evidenced and the influence of the embedded ligand is highlighted, thus allowing proposal of a mechanism for the hydrogen evolution reaction catalyzed by these clusters.
Sigma-Aldrich
Terephthalic acid, 98%