Merck
  • Alkyne-tag Raman imaging for visualization of mobile small molecules in live cells.
Alkyne-tag Raman imaging for visualization of mobile small molecules in live cells.
Journal of the American Chemical Society (2012-12-04)
Hiroyuki Yamakoshi, Kosuke Dodo, Almar Palonpon, Jun Ando, Katsumasa Fujita, Satoshi Kawata, Mikiko Sodeoka
ABSTRACT
Alkyne has a unique Raman band that does not overlap with Raman scattering from any endogenous molecule in live cells. Here, we show that alkyne-tag Raman imaging (ATRI) is a promising approach for visualizing nonimmobilized small molecules in live cells. An examination of structure-Raman shift/intensity relationships revealed that alkynes conjugated to an aromatic ring and/or to a second alkyne (conjugated diynes) have strong Raman signals in the cellular silent region and can be excellent tags. Using these design guidelines, we synthesized and imaged a series of alkyne-tagged coenzyme Q (CoQ) analogues in live cells. Cellular concentrations of diyne-tagged CoQ analogues could be semiquantitatively estimated. Finally, simultaneous imaging of two small molecules, 5-ethynyl-2'-deoxyuridine (EdU) and a CoQ analogue, with distinct Raman tags was demonstrated.
Sigma-Aldrich
2′-Deoxyuridine, ≥98.5%