Merck
  • High-specificity single-tube multiplex genotyping using Ribo-PAP PCR, tag primers, alkali cleavage of RNA/DNA chimeras and MALDI-TOF MS.
High-specificity single-tube multiplex genotyping using Ribo-PAP PCR, tag primers, alkali cleavage of RNA/DNA chimeras and MALDI-TOF MS.
Human mutation (2012-11-08)
Florence Mauger, David H Gelfand, Amar Gupta, Veeraiah Bodepudi, Stephen G Will, Keith Bauer, Thomas W Myers, Ivo G Gut
ABSTRACT
Here, we describe a high-throughput, single-tube, allele-specific ribonucleotide analog pyrophosphorolysis-activated polymerization (ribo-PAP) PCR multiplex genotyping and resequencing method. An RNA/DNA chimeric PCR product is generated using genomic DNA as starting template, a panel of allele-selective 5'-tagged primers, a reverse primer, one nucleotide in the ribo-form (90-100%), the other nucleotides in the deoxy-form, a DNA polymerase capable of incorporating ribonucleotides, a suitable buffer and thermal cycling. The RNA/DNA chimeric PCR products are fragmented by treatment with alkali and analyzed by mass spectrometry. All allele-selective primers have a 5' repetitive motif where each repeat unit has a unique, distinct mass upon reverse copying and alkali fragmentation. The mass of the complement repeat fragment or flag identifies the primer or primers that were recruited in the ribo-PAP PCR. The method readily identifies homozygous and heterozygous positions in simplex or duplex ribo-PAP PCR. Many different tags can be analyzed simultaneously. The assay can genotype several SNPs in a single tube. It thus constitutes the simplest genotyping protocol with multiplex analysis. This novel genotyping and resequencing protocol was applied to different genomic loci: NOS1 and H19 in 30 individuals in simplex ribo-PAP PCR and at two SLCO1B1 loci in 95 individuals in duplex ribo-PAP PCR.
Sigma-Aldrich
Pyrophosphoric acid, technical grade
Pricing and availability is not currently available.