Merck
  • Effective Friedel-Crafts acylation of biotin acid chloride in trifluoromethanesulfonic acid.
Effective Friedel-Crafts acylation of biotin acid chloride in trifluoromethanesulfonic acid.
Bioscience, biotechnology, and biochemistry (2012-11-08)
Yasuyuki Muto, Yuta Murai, Yasuko Sakihama, Yasuyuki Hashidoko, Makoto Hashimoto
ABSTRACT
Biotin is one of the most useful tags in (bio)analytical science due to its specific interaction with avidin, but is not easy to convert because of its low solubility in most solvents. Friedel-Crafts acylation of biotin acid chloride in triflic acid was examined, and the synthesized derivatives had stronger affinity to avidin than biotin in a binding assay using 2-(4'-hydroxyphenylazo)benzoic acid.
Sigma-Aldrich
Lanthanum(III) trifluoromethanesulfonate, 99.999% trace metals basis
Sigma-Aldrich
Zinc trifluoromethanesulfonate, 98%
Sigma-Aldrich
Trifluoromethanesulfonic acid, ReagentPlus®, ≥99%
Sigma-Aldrich
Lithium trifluoromethanesulfonate, 99.995% trace metals basis
Sigma-Aldrich
Trifluoromethanesulfonic acid, reagent grade, 98%
Sigma-Aldrich
Lithium trifluoromethanesulfonate, 96%
Sigma-Aldrich
Aluminum trifluoromethanesulfonate, 99.9% trace metals basis
Sigma-Aldrich
Silver trifluoromethanesulfonate, ≥99%
Sigma-Aldrich
Silver trifluoromethanesulfonate, purum, ≥98.0% (Ag)
Sigma-Aldrich
Silver trifluoromethanesulfonate, ≥99.95% trace metals basis