Merck
  • Transamidation of primary amides with amines catalyzed by zirconocene dichloride.
Transamidation of primary amides with amines catalyzed by zirconocene dichloride.
Chemical communications (Cambridge, England) (2012-10-26)
Benjamin N Atkinson, A Rosie Chhatwal, Helen V Lomax, James W Walton, Jonathan M J Williams
ABSTRACT
Zirconocene dichloride (Cp(2)ZrCl(2)) has been shown to be an effective catalyst for the transamidation of primary amides with amines in cyclohexane at 80 °C in 5-24 hours. For favourable substrates, the reaction can be performed at temperatures as low as 30 °C.
Sigma-Aldrich
Bis(cyclopentadienyl)zirconium(IV) dichloride, ≥98%