Merck
Bright core-shell semiconductor quantum wires.
Journal of the American Chemical Society (2012-10-26)
Yi-Hsin Liu, Fudong Wang, Jessica Hoy, Virginia L Wayman, Lindsey K Steinberg, Richard A Loomis, William E Buhro
ABSTRACT
Colloidal CdTe quantum wires are reported having ensemble photoluminescence efficiencies as high as 25% under low excitation-power densities. High photoluminescence efficiencies are achieved by formation of a monolayer CdS shell on the CdTe quantum wires. Like other semiconductor nanowires, the CdTe quantum wires may contain frequent wurtzite-zinc-blende structural alternations along their lengths. The present results demonstrate that the optical properties, emission-peak shape and photoluminescence efficiencies, are independent of the presence or absence of such structural alternations.