Merck
  • Comprehensive evaluation of the effects of enalapril on matrix metalloproteinases levels in hypertension.
Comprehensive evaluation of the effects of enalapril on matrix metalloproteinases levels in hypertension.
Cardiovascular drugs and therapy (2012-10-23)
Vanessa Fontana, Pamela S Silva, Tatiane C Izidoro-Toledo, Celso Biagi, Eduardo B Oliveira, Raquel F Gerlach, Jose E Tanus-Santos
ABSTRACT
Angiotensin-converting enzyme inhibitors (ACEi) may downregulate matrix metalloproteinases (MMPs). We examined whether enalapril affects MMP-2, MMP-8, and MMP-9 levels and activity, and their endogenous inhibitors (tissue inhibitors of MMPs, TIMP-1 and TIMP-2) levels in hypertensive patients. Moreover, we assessed the effects of enalaprilat on MMP-9 and TIMP-1 secretion by human endothelial cells (HUVECs). Thirty-eight hypertensive patients received enalapril for 8 weeks and were compared with thirty-eight normotensive controls. Blood samples were collected at baseline and after treatment. Plasma ACE activity was determined by a fluorimetric assay. Plasma MMP-2, MMP-8, MMP-9, TIMP-1, and TIMP-2 were measured by ELISA and gelatin zymography. A fluorogenic peptide cleavage assay was used to measure MMP activity. HUVECs cells were stimulated by phorbol-12-myristate-13-acetate (PMA) and the effects of enalaprilat (10(-10) to 10(-6) M) on MMP-9 and TIMP-1 levels were determined. Enalapril decreased blood pressure and ACE activity in hypertensive patients (P < 0.05), but had no effects on plasma MMP-2, MMP-8, MMP-9, TIMP-1, and TIMP-2 levels, or MMP activity. Enalaprilat had no effects on PMA-induced increases in MMP-9 and TIMP-1 secretion by HUVECs or on MMP activity. We show consistent evidence, both in vivo and in vitro, that enalapril does not affect MMPs and TIMPs levels in hypertensive patients.
Sigma-Aldrich
Enalaprilat dihydrate, ≥98% (HPLC)