Merck
  • Novel bio-active lipid nanocarriers for the stabilization and sustained release of sitosterol.
Novel bio-active lipid nanocarriers for the stabilization and sustained release of sitosterol.
Nanotechnology (2012-10-16)
I Lacatusu, N Badea, R Stan, A Meghea
ABSTRACT
In this work, new stable and efficiently bio-active lipid nanocarriers (NLCs) with antioxidant properties have been developed for the transport of active ingredients in food. The novel NLCs loaded with β-sitosterol/β-sitosterol and green tea extract (GTE) and prepared by a combination of natural oils (grape seed oil, fish oil and squalene) and biological lipids with food grade surfactants, were physico-chemically examined by DLS, TEM, electrokinetic potential, DSC and HPLC and found to have main diameters less than 200 nm, a spherical morphology, excellent physical stability, an imperfect crystalline lattice and high entrapment efficiency. The novel loaded-NLCs have demonstrated the potential to develop a high blocking action of chain reactions, trapping up to 92% of the free-oxygen radicals, as compared to the native β-sitosterol (AA%=36.5). Another advantage of this study is associated with the quality of bio-active NLCs based on grape seed oil and squalene to manifest a better sitosterol-sustained release behaviour as compared to their related nanoemulsions. By coupling both in vitro results, i.e. the enhanced antioxidant activity and superior release properties, this study emphasizes the sustainability of novel bio-active nanocarriers to gain specific bio-food features for development of functional foods with a high applicability spectrum.
Sigma-Aldrich
β-Sitosterol, ≥70%
Supelco
β-Sitosterol, certified reference material, 100 μg/mL in chloroform
Sigma-Aldrich
β-Sitosterol, synthetic, ≥95%
Sigma-Aldrich
β-Sitosterol, from soybean, ≥96%