Merck
  • Biogenic phosphorus compounds in sediment and suspended particles in a shallow eutrophic lake: a ³¹P-nuclear magnetic resonance (³¹P NMR) study.
Biogenic phosphorus compounds in sediment and suspended particles in a shallow eutrophic lake: a ³¹P-nuclear magnetic resonance (³¹P NMR) study.
Environmental science & technology (2012-09-22)
Ryuichiro Shinohara, Akio Imai, Nobuyuki Kawasaki, Kazuhiro Komatsu, Ayato Kohzu, Shingo Miura, Tomoharu Sano, Takayuki Satou, Noriko Tomioka
ABSTRACT
Differences in biogenic phosphorus (P) compounds between sediment and suspended particles in aquatic environments are important for understanding the mechanisms of internal P loading, but these differences are still unknown. We used solution-state (31)P-nuclear magnetic resonance spectroscopy ((31)P NMR) with NaOH-ethylenediaminetetraacetic extraction to detect the multiple P compounds in suspended particles and sediment in the eutrophic Lake Kasumigaura, including orthophosphate monoesters, orthophosphate diesters, pyrophosphate, and polyphosphate. We tested the hypothesis that there is a significant difference between these groups in suspended particles and sediment. Biogenic P other than orthophosphate was found in significantly higher proportions in suspended particles (74.3% of total P) than in sediment (25.6%). Orthophosphate monoesters were comparatively more abundant in suspended particles, as indicated by the ratio of orthophosphate diesters to monoesters (average, 0.31 for suspended particles; 1.05 for sediment). The compounds identified as orthophosphate monoesters by (31)P NMR spectroscopy originated mainly from phospholipids (α-glycerophosphate and β-glycerophosphate) and ribonucleic acid (RNA-P), whereas the orthophosphate diesters included mostly DNA (DNA-P). These results suggest that the dynamics of orthophosphate diesters, the production of DNA-P, or the degradation of phospholipids, play an important role in P cycling in Lake Kasumigaura.
Sigma-Aldrich
DL-α-Glycerol phosphate magnesium salt hydrate, ~85% (KT)