Merck
  • Optimisation for subcritical fluid extraction of 17-methyltestosterone with 1,1,1,2-tetrafluoroethane for HPLC analysis.
Optimisation for subcritical fluid extraction of 17-methyltestosterone with 1,1,1,2-tetrafluoroethane for HPLC analysis.
Food chemistry (2012-09-18)
Yuqian Han, Qinchuan Ma, Jie Lu, Yong Xue, Changhu Xue
ABSTRACT
A simple, rapid and sensitive method was developed for determination of 17α-methyltestosterone in aquatic products by extraction with subcritical 1,1,1,2-tetrafluoroethane (R134a) extraction and high performance liquid chromatography (HPLC). Response surface methodology (RSM) was adopted to optimise extraction pressure, temperature and co-solvent volume. The optimum extraction conditions predicted within the experimental ranges were as follows: pressure 5 MPa, temperature 31°C, and co-solvent volume 3.35ml. The analysis was carried out on XDB-C(18) column (4.6 mm × 250 mm, 5 μm) with the mobile phase acetonitrile-water (55:45, v/v), flow rate 0.8 ml/min, temperature 30°C and wavelength 245 nm. Good linearity of detection was obtained for 17α-methyltestosterone between concentrations of 50-250 ng/ml, r(2)=0.999. The method was validated using samples fortified with 17α-methyltestosterone at levels of 10, 30 and 50 ng/g, the mean recovery exceeds 90%, and the RSD values were less than 10%.
Sigma-Aldrich
17α-Methyltestosterone, ≥97.0% (HPLC)
Supelco
17α-Methyltestosterone, VETRANAL®, analytical standard
Sigma-Aldrich
17α-Methyltestosterone, solid (photosensitive)
Supelco
17α-Methyltestosterone solution, 1.0 mg/mL in 1,2-dimethoxyethane, ampule of 1 mL, certified reference material, Cerilliant®