Merck
  • Non-enzymatic conversion of chlorophyll-a into chlorophyll-d in vitro: a model oxidation pathway for chlorophyll-d biosynthesis.
Non-enzymatic conversion of chlorophyll-a into chlorophyll-d in vitro: a model oxidation pathway for chlorophyll-d biosynthesis.
FEBS letters (2012-06-05)
Takanori Fukusumi, Kohei Matsuda, Tadashi Mizoguchi, Tomohiro Miyatake, Satoshi Ito, Tsukasa Ikeda, Hitoshi Tamiaki, Toru Oba
ABSTRACT
Chlorophyll-a (Chl-a) was readily converted into Chl-d under mild conditions without any enzymes. Treatment of Chl-a dissolved in dry tetrahydrofuran (THF) with thiophenol and acetic acid at room temperature successfully produced Chl-d in 31% yield. During the acidic oxidation, removal of the central magnesium, pheophytinization, was sufficiently suppressed. This mild pathway can give insights into the yet unidentified Chl-d biosynthesis.
Sigma-Aldrich
Thiophenol, 97%
Sigma-Aldrich
Sodium thiophenolate, technical grade, 90%
Sigma-Aldrich
Thiophenol, ≥99%
Supelco
Thiophenol, analytical standard