Merck
  • Enantiopure synthesis of side chain-modified α-amino acids and 5-cis-alkylprolines.
Enantiopure synthesis of side chain-modified α-amino acids and 5-cis-alkylprolines.
The Journal of organic chemistry (2012-05-24)
Amar R Mohite, Ramakrishna G Bhat
ABSTRACT
A short, concise synthesis of enantiopure, side chain-modified α-amino acids such as 4-oxo-L-norvaline, 6-oxo-L-homonorleucine, and 5-cis-alkyl prolines is described. Knoevenagel condensation of l-aminocarboxylate-derived β-ketoesters with aldehydes followed by reductive decarboxylation results in unnatural α-amino acids in good yield. A fluorescent amino acid is synthesized using a similar protocol. These studies show that aminocarboxylate-derived β-ketoesters are very useful intermediates and the method employed is both general and practical for the preparation of γ(δ)-oxo α-amino acids and alkylprolines.
Sigma-Aldrich
L-Norleucine, ≥98% (TLC)
Sigma-Aldrich
L-Norleucine, suitable for amino acid analysis, BioReagent