Merck
Fragrance material review on ethyl phenyl carbinyl acetate.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association (2012-03-22)
D McGinty, C S Letizia, A M Api
ABSTRACT
A toxicologic and dermatologic review of ethyl phenyl carbinyl acetate when used as a fragrance ingredient is presented. Ethyl phenyl carbinyl acetate is a member of the fragrance structural group Aryl Alkyl Alcohol Simple Acid Esters (AAASAE). The AAASAE fragrance ingredients are prepared by reacting an aryl alkyl alcohol with a simple carboxylic acid (a chain of 1-4 carbons) to generate formate, acetate, propionate, butyrate, isobutyrate and carbonate esters. This review contains a detailed summary of all available toxicology and dermatology papers that are related to this individual fragrance ingredient and is not intended as a stand-alone document. Available data for ethyl phenyl carbinyl acetate were evaluated, then summarized, and includes: physical properties; acute toxicity; skin irritation; and skin sensitization data. A safety assessment of the entire AAASAE will be published simultaneously with this document; please refer to Belsito et al. (2012) for an overall assessment of the safe use of this material and all AAASAE in fragrances.
Sigma-Aldrich
Methyl phenylcarbinyl acetate, ≥98%, FCC, FG