Merck
  • In vivo measurements of blood flow in a rat using X-ray imaging technique.
In vivo measurements of blood flow in a rat using X-ray imaging technique.
The international journal of cardiovascular imaging (2012-02-23)
Sung Yong Jung, Sungsook Ahn, Kweon Ho Nam, Jin Pyung Lee, Sang Joon Lee
ABSTRACT
To measure instantaneous velocity fields of venous blood flow in a rat using X-ray particle tracking method. Gold nanoparticles (AuNPs) incorporated chitosan microparticles were applied as biocompatible flow tracers. After intravenous injection of the AuNP-chitosan particles into 7- to 9-week-old male rat vein, X-ray images of particle movement inside the cranial vena cava were consecutively captured. Individual AuNP-chitosan particles in the venous blood flow were clearly observed, and the corresponding velocity vectors were successfully extracted. The measured velocity vectors are in good agreement with the theoretical velocity profile suggested by Casson. This is the first trial to measure blood flow in animals under in vivo conditions with X-ray imaging technique. The results show that X-ray particle tracking technique has a great potential for in vivo measurements of blood flow, which can extend to various biomedical applications related with the diagnosis of circulatory vascular diseases.
Sigma-Aldrich
Sodium tetrachloroaurate(III) dihydrate, 99%
Sigma-Aldrich
Gold(III) chloride solution, 99.99% trace metals basis, 30 wt. % in dilute HCl
Sigma-Aldrich
Potassium gold(III) chloride, 98%
Sigma-Aldrich
Potassium gold(III) chloride, 99.995% trace metals basis