Merck
  • Stability studies of clonazepam, diazepam, haloperidol, and doxepin with diverse polarities in an acidic environment.
Stability studies of clonazepam, diazepam, haloperidol, and doxepin with diverse polarities in an acidic environment.
Journal of AOAC International (2012-02-11)
Anna Maślanka, Jan Krzek, Mariusz Stolarczyk, Maria Walczak, Agata Głogowska
ABSTRACT
Stability of clonazepam, diazepam, haloperidol, and doxepin was determined in acidic solutions. In addition, determination of the kinetic and thermodynamic properties of this stability was carried out. Reaction rate constants (k), half-life times (t(0.1) and t(0.5)), and activation energy (Ea) were estimated for the drugs, which differed in polarity expressed with log P values. It was observed that estimated Ea values increased from 42.13 to 125.03 kJ/mol with an increase of lipophilicity (log P) beginning from the most hydrophilic drug (clonazepam, 2.70 log P) to the most lipophilic drug (doxepin, 4.10 log P). All degradation products were studied using an HPLC/electrospray ionization-MS technique in the positive ionization mode.
Product Description
Doxepin for system suitability, European Pharmacopoeia (EP) Reference Standard
Supelco
Doxepin hydrochloride solution, 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Doxepin hydrochloride, ~85% E-isomer basis, ≥98% (GC), 15% Z-isomer basis, powder