Merck
  • A highly active and magnetically retrievable nanoferrite-DOPA-copper catalyst for the coupling of thiophenols with aryl halides.
A highly active and magnetically retrievable nanoferrite-DOPA-copper catalyst for the coupling of thiophenols with aryl halides.
Chemical communications (Cambridge, England) (2012-02-02)
R B Nasir Baig, Rajender S Varma
ABSTRACT
Diaryl sulfides were synthesized using a magnetically recoverable heterogeneous Cu catalyst via one-pot multi component reaction using MW irradiation; the use of isopropanol as a benign reaction medium, easy recovery of the catalyst using an external magnet, efficient recycling, and the high stability of the catalyst render the protocol economic and sustainable.
Sigma-Aldrich
Thiophenol, 97%
Sigma-Aldrich
Sodium thiophenolate, technical grade, 90%
Supelco
Thiophenol, analytical standard
Sigma-Aldrich
Chlorobenzene, anhydrous, 99.8%
Supelco
Chlorobenzene, analytical standard
Sigma-Aldrich
Thiophenol, ≥99%
Sigma-Aldrich
Chlorobenzene, ACS reagent, ≥99.5%
Sigma-Aldrich
Chlorobenzene, suitable for HPLC, 99.9%