Merck
  • Diversity oriented syntheses of conventional heterocycles by smart multi component reactions (MCRs) of the last decade.
Diversity oriented syntheses of conventional heterocycles by smart multi component reactions (MCRs) of the last decade.
Molecules (Basel, Switzerland) (2012-01-24)
Heiner Eckert
ABSTRACT
A collection of smart multicomponent reactions (MCRs) with continuative post condensation cyclizations (PCCs) is presented to construct conventional three- to seven-membered heterocyclic compounds in diversity oriented syntheses (DOS). These will provide a high degree of applying economical and ecological advantages as well as of practicability. Water, ionic liquids, and solvent-less syntheses as well as use of various forms of energy as microwave and ultrasonic irradiation are examined and discussed.
Sigma-Aldrich
Acetylenedicarboxylic acid monopotassium salt, ≥98%
Sigma-Aldrich
Acetylenedicarboxylic acid, 95%