Merck
  • Transferability of a modified embryonic stem cell test using a new endpoint for developmental neurotoxicity.
Transferability of a modified embryonic stem cell test using a new endpoint for developmental neurotoxicity.
Toxicology mechanisms and methods (2011-11-16)
Dae Hyun Baek, Su-Yeon An, Jae Hyun Park, Youngju Choi, Ki Dae Park, Jin Wook Kang, Kyoung Suk Choi, Sung Hee Park, Min Young Whang, Jiyou Han, Jong-Hoon Kim, Hyung Soo Kim, Dongho Geum, Tae Moo Yoo
ABSTRACT
We developed and analyzed a new surrogate endpoint of the mouse embryonic stem cell test (EST) for developmental neurotoxicity. To determine the sensitivity, specificity, and transferability of the new endpoint, a pre-validation team from three independent laboratories optimized and standardized the protocol for neuronal differentiation of mouse embryonic stem cells (mESCs) by measuring the neuronal differentiation rates of mESCs under different culture conditions, such as the presence or absence of basic fibroblast growth factor (bFGF) in the growth media and varying lengths of culture. In addition, a component ratio of neuronal cells was measured by using flow cytometry analysis of β-III tubulin (Tuj1)-positive cells and real-time polymerase chain reaction analysis of microtubule-associated protein 2 (MAP2) mRNA. Our results showed that the best growth was achieved by culturing mESCs for 12 d in N2B27 medium without bFGF or ascorbic acid. Lead (II) acetate and aroclor 1254 were used to test the usefulness of the new endpoint. When we used the known ID(50) values for lead (II) acetate in the EST model, it was classified as non-embryotoxic; however, when we used the new ID(50) values that we determined in this study, it was classified as weakly embryotoxic. Aroclor 1254 and penicillin G were also classified as weakly embryotoxic and non-embryotoxic compounds, respectively, when cardiac and neuronal differentiation ID(50) values were used. Therefore, our new surrogate endpoint for developmental neurotoxicity is not only sensitive and specific but also transferable among laboratories.
Product Description
Supelco
Aroclor 1254 solution, certified reference material, 500 mg/kg in transformer oil
Supelco
Aroclor 1254, ampule of 50 mg, analytical standard
Supelco
Aroclor 1254 solution, certified reference material, 50 mg/kg in transformer oil