- [Study on the chemical constituents of Pithecellobium clypearia].
[Study on the chemical constituents of Pithecellobium clypearia].
Zhong yao cai = Zhongyaocai = Journal of Chinese medicinal materials (2011-11-10)
Chun-ying Xie, Le-wei Lin
PMID22066399
ABSTRACT
To study the chemical constituents of Pithecellobium clypearia. Silica gel column chromatography and Sephadex LH-20 were used to separate and purify compounds from the EtOAc soluble fraction of Pithecellobium clypearia. 9 compounds were elucidated on the basis of physicochemical properties and spectrascopic analysis. They were identified as beta-Sitosterol (1), tritriacontane (2), 5-hydroxy-3,7,3',4'-tetramethoxyflavone (3), oleanolic acid (4), 5,4'-dihydroxy-3,7,3'-trimethoxyflavone (5), alpha-amyrin (6), luteolin (7), ursolic acid (8), luteoloside (9). Compounds 3 and 5 are isolated from this plant for the first time.