Merck
  • MCM-41-supported oxo-vanadium(IV) complex: a highly selective heterogeneous catalyst for the bromination of hydroxy aromatic compounds in water.
MCM-41-supported oxo-vanadium(IV) complex: a highly selective heterogeneous catalyst for the bromination of hydroxy aromatic compounds in water.
Langmuir : the ACS journal of surfaces and colloids (2011-10-27)
Susmita Bhunia, Debraj Saha, Subratanath Koner
ABSTRACT
An ecofriendly solid catalyst has been synthesized by anchoring vanadium(IV) into organically modified MCM-41. First, the surface of Si-MCM-41 was modified with 3-aminopropyl-triethoxysilane (3-APTES), the amine group of which upon condensation with ortho-hydroxy-acetophenone affords a N(2)O(2)-type Schiff base moiety in the mesoporous matrix. The Schiff base moieties were used to anchor oxo-vanadium(IV) ions. The prepared catalyst has been characterized by UV-vis, IR spectroscopy, small-angle X-ray diffraction (SAX), nitrogen sorption, and transmission electron microscopy (TEM) studies. It is observed that the mesostructure has not been destroyed in the multistep synthesis procedure, as evidenced by SAX and TEM measurements. The catalyst has shown unprecedented high conversion as well as para selectivity toward the bromination of hydroxy aromatic compounds using aqueous 30% H(2)O(2)/KBr in water. The reaction proceeds according to the stoichiometric ratio, and the monobrominated product was obtained as the major product using a stoichiometric amount of the bromine source. The immobilized complex does not leach or decompose during the catalytic reactions, showing practical advantages over the free metal complex.
Sigma-Aldrich
Bromophenol Blue, titration: suitable
Sigma-Aldrich
Phenol Red, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Phenol red solution, 0.5%, liquid, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Phenol Red, ACS reagent
Sigma-Aldrich
Bromophenol Blue, ACS reagent
Sigma-Aldrich
Potassium bromide, anhydrous, powder, 99.999% trace metals basis
Sigma-Aldrich
Potassium bromide, BioUltra, ≥99.5% (AT)
Sigma-Aldrich
Potassium bromide, anhydrous, powder, 99.95% trace metals basis
Sigma-Aldrich
Potassium bromide, BioXtra, ≥99.0%
Sigma-Aldrich
Potassium bromide, ACS reagent, ≥99.0%
Sigma-Aldrich
Potassium bromide, ≥99% trace metals basis
Sigma-Aldrich
Potassium bromide, ReagentPlus®, ≥99.0%