Merck
  • Conformational analysis of 18-azacrown-6 and its bonding with late first transition series divalent metals: insight from DFT combined with NPA and QTAIM analyses.
Conformational analysis of 18-azacrown-6 and its bonding with late first transition series divalent metals: insight from DFT combined with NPA and QTAIM analyses.
The journal of physical chemistry. A (2011-10-04)
Pradeep R Varadwaj, Arpita Varadwaj, Gilles H Peslherbe, Helder M Marques
ABSTRACT
Density functional theory calculations, together with quantum theory of atoms in molecules (QTAIM) analyses, have been performed to investigate 18-azacrown-6 complexes of the high-spin late first transition series divalent metal ions in the gas phase and, in some cases, in aqueous solution simulated by a polarizable continuum model. Six intramolecular H-H bonding interactions in the meso-complexes are found to arise from folding of the ligand upon its electrostatic interaction with the metal ions, which are largely absent in the lowest-energy C(2h) conformer of the free ligand. The ligand-to-metal charge transfer obtained from QTAIM analysis, among other things, is found to be an important factor that controls the stability of these complexes. The inter-relationship between the ligand preorganization energy, the zero-point corrected formation energy of the metal complexes, and the H-H bonding pair distances, as well as the dependence of the electron density and the total energy density at the H-H bond critical points on the H-H bonding pair distances, provides a physical basis for understanding and explaining the stabilizing nature of these closed-shell interactions, which are often viewed as steric clashes that lead to complex destabilization.
Sigma-Aldrich
18-Crown-6, ≥99.0%
Sigma-Aldrich
18-Crown-6, 99%
Sigma-Aldrich
18-Crown-6, purum, ≥99.0% (GC)
Sigma-Aldrich
18-Crown-6 solution, 1.0 M in THF