Merck
  • Comparison of molecular dynamics simulation methods for amyloid β(1-42) monomers containing D-aspartic acid residues for predicting retention times in chromatography.
Comparison of molecular dynamics simulation methods for amyloid β(1-42) monomers containing D-aspartic acid residues for predicting retention times in chromatography.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2011-08-30)
Akifumi Oda, Kana Kobayashi, Ohgi Takahashi
ABSTRACT
Molecular dynamics simulations of amyloid β(1-42) containing D-aspartic acid residues were performed using several continuous solvent models to investigate the usefulness of simulation methods for D-amino acid-containing proteins and peptides. Normal molecular dynamics simulations and replica exchange molecular dynamics simulations, which are one of the generalized-ensemble algorithms, were performed. Because the β-structure contents of amyloid β(1-42) peptides obtained by replica exchange molecular dynamics simulations with Onufriev-Bashford-Case generalized Born implicit solvent were qualitatively consistent with experimental data, replica exchange molecular dynamics rather than other methods appeared to be more reasonable for calculations of amyloid β(1-42) containing D-aspartic acid residues. Computational results revealed that peptides with stereoinversion of Asp23 tend to form β-sheet structures by themselves, in contrast to the wild-type peptides that form β-sheet structures only after aggregation. These results are expected to be useful for computational investigations of proteins and peptides such as prediction of retention time of peptides and proteins containing D-aspartic acid residues.
Sigma-Aldrich
D-Aspartic acid, ReagentPlus®, 99%