Merck
  • Comparative study of 6-APA production by free and agar immobilized bacteria in nutrient broth culture.
Comparative study of 6-APA production by free and agar immobilized bacteria in nutrient broth culture.
Indian journal of experimental biology (2011-05-28)
A K Dolui, S Das
ABSTRACT
In the present study different bacterial samples were isolated from soil of different places of Dibrugarh and screened for biotransformation ability to produce 6-Aminopenicillanic acid. Among ten isolated bacterial samples, three gram positive bacterial samples designated as AKDD-2, AKDD-4 and AKDD-6 showed the production of 6-APA from penicillin G. Assessment of production of 6-APA after incubation in penicillin G (2 mg/ml) by three different samples separately in free and agar immobilization state was done by HPLC analysis. Reusability of immobilized cells was found successful up to 14 days.
Sigma-Aldrich
(+)-6-Aminopenicillanic acid, 96%