Merck
  • Sonochemical syntheses of a new nano-plate cadmium(II) coordination polymer as a precursor for the synthesis of cadmium(II) oxide nanoparticles.
Sonochemical syntheses of a new nano-plate cadmium(II) coordination polymer as a precursor for the synthesis of cadmium(II) oxide nanoparticles.
Ultrasonics sonochemistry (2011-01-29)
Maryam Ramazani, Ali Morsali
ABSTRACT
Nanoplates of the three-dimensional coordination polymer, {[Cd(3)(3-pyc)(4)(N(3))(2)(H(2)O)](n) (1), 3-pyc(-)=pyridine-3-carboxylate), have been synthesized by a sonochemical process and characterized by scanning electron microscopy, X-ray powder diffraction, IR spectroscopy and elemental analyses. Cadmium(II) oxide nanoparticles were prepared from thermal decomposition in oleic acid and direct calcination of compound 1 at different temperatures. The thermal stability of nano-sized compound 1 was studied by thermal gravimetric (TG) and differential thermal analyses (DTA). Results show that the size and morphology of the CdO nanoparticles are dependent upon the particles size of compound 1 and the thermolysis temperature. A decrease in the particle size of compound 1 leads to a decrease in the particle size of the CdO, while an increase in the processing temperature leads to an increase in the particle size of the produced cadmium(II) oxide nano-particles.
Sigma-Aldrich
Cadmium oxide, powder, 99.5% trace metals basis
Sigma-Aldrich
Cadmium oxide, ≥99.99% trace metals basis