Merck
  • ¹H nuclear magnetic resonance based metabolic urinary profiling of patients with ischemic heart failure.
¹H nuclear magnetic resonance based metabolic urinary profiling of patients with ischemic heart failure.
Clinical biochemistry (2010-12-21)
Seok-Min Kang, Jong-Chul Park, Min-Jeong Shin, Hyeran Lee, Jaewon Oh, Do Hyun Ryu, Geum-Sook Hwang, Ji Hyung Chung
ABSTRACT
We sought to identify metabolic pathways characterizing human heart failure (HF) using ¹NMR based urinary metabolomic analysis in conjunction with multivariate statistics. Patients with systolic HF of ischemic origin (n=15) and healthy controls (n=20) participated in this study. Patients with type 2 diabetes mellitus were excluded. The results showed that the urine of the HF patients had higher levels of metabolites for acetate (p<0.05) and acetone (p<0.01) compared to the healthy controls. In addition, there was a perturbation in methylmalonate metabolism as shown by increased urinary levels of methylmalonic acid (p<0.001) in the HF patients. HF patients also had increased urinary levels of cytosine (p<0.01) and phenylacetylglycine (p<0.01) and decreased 1-methylnicotinamide (p<0.05) compared to healthy controls. TCA cycle metabolites and fatty acid metabolism were modified in the HF patients, indicating altered energy metabolism. Moreover, perturbations of metabolism in nucleotide and methylmalonate were observed.
Sigma-Aldrich
L-Alanine, ≥99%
Sigma-Aldrich
L-Alanine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Leucine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Leucine, 99%, FG
Sigma-Aldrich
L-Alanine, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Alanine, ≥98% (TLC)
Sigma-Aldrich
L-Leucine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Threonine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 99.0-101.0%
Sigma-Aldrich
L-Leucine, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
L-Threonine, reagent grade, ≥98% (HPLC)
Supelco
L-Leucine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
SAFC
L-Threonine
Sigma-Aldrich
L-Threonine, BioXtra, ≥99.5% (NT)
Supelco
L-Threonine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
L-Alanine, Pharmaceutical Secondary Standard; Certified Reference Material
Alanine, European Pharmacopoeia (EP) Reference Standard
Supelco
L-Alanine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
L-Leucine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Threonine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Isoleucine, 99%, FCC, FG
Sigma-Aldrich
L-Isoleucine, BioUltra, ≥99.5% (NT)
Supelco
L-Serine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Glutamic acid, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Methionine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
Taurine, BioUltra, ≥99.5% (T)
SAFC
Taurine
Sigma-Aldrich
L-Glutamic acid, FCC
Sigma-Aldrich
Taurine, suitable for cell culture, meets USP testing specifications
Sigma-Aldrich
Taurine, ≥99%
Sigma-Aldrich
L-Serine, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 98.5-101.0%