Merck
  • Deep anesthesia suppresses ventricular tachyarrhythmias in rabbit model of the acquired long QT syndrome.
Deep anesthesia suppresses ventricular tachyarrhythmias in rabbit model of the acquired long QT syndrome.
Circulation journal : official journal of the Japanese Circulation Society (2010-12-09)
Hideko Inaba, Noriyuki Hayami, Kosuke Ajiki, Tomoyuki Kunishima, Hidenori Watanabe, Kenta Tsutsui, Noboru Yamagishi, Satoshi Yamagishi, Anna Sugiura, Takashi Mikamo, Yuji Murakawa
ABSTRACT
Anesthesia sometimes suppresses ventricular tachyarrhythmias (VT) resistant to conventional pharmacological treatment. To know (1) whether deep anesthesia inhibits abnormal repolarization-related VT and (2) if α2-adrenoreceptor (AR) agonistic action is associated with the antiarrhythmic effect of anesthetics, the incidence of VT in a rabbit model of acquired long QT syndrome using different anesthetic regimen was assessed. In Study 1 (n = 30), 15 rabbits were lightly anesthetized with ketamine (123 ± 46 mg/kg) and an α2-AR agonist, xylazine (9.4±3.0mg/kg), while combination of these anesthetics at high doses were used in the other 15 rabbits (343 ± 78 mg/kg and 38.9 ± 3.0 mg/kg). Administration of α1-AR stimulant, methoxamine and nifekalant (Ikr blocker) caused VT in all lightly anesthetized rabbits. In contrast, VT was observed only in 1 of the 15 deeply anesthetized rabbits (P < 0.01). In Study 2 (n = 15), 10 rabbits were anesthetized with high-dose ketamine and low-dose xylazine. In the other 5 rabbits, low-dose ketamine and high-dose xylazine were used. QTc interval in the latter was longer than that of the former (399 ± 56 ms vs. 494 ± 57 ms, P < 0.01). Although no VT appeared in high/low-rabbits, VT occurred in 3 out of 5 low/high-rabbits (P < 0.05). These results suggest that (1) deep anesthesia suppresses abnormal repolarization-related VT and (2) antiarrhythmic effect of anesthesia on this type of VT is not dependent on α2-AR agonistic action.
Sigma-Aldrich
Methoxamine hydrochloride