Merck
  • Synthesis of a D-Ala-D-Ala peptide isostere via olefin cross-metathesis and evaluation of vancomycin binding.
Synthesis of a D-Ala-D-Ala peptide isostere via olefin cross-metathesis and evaluation of vancomycin binding.
Bioorganic & medicinal chemistry letters (2010-07-03)
Ryan K Quinn, Amelia L Cianci, Jennifer A Beaudoin, Bianca R Sculimbrene
ABSTRACT
The alkene peptide isostere for the d-Ala-d-Ala dipeptide was synthesized via a convergent approach utilizing olefin cross-metathesis. The new isostere was then evaluated for binding to the last resort antibiotic, vancomycin. The alkene isostere exhibited a K(D)=90 microM in comparison to the native peptide (K(D)=2.3 microM) and Lac mutant (K(D)=2300 microM). This study demonstrates that loss of binding in vancomycin resistant strains as a result of a d-Ala to d-Lac mutation is from both the loss of a crucial hydrogen bond and introduction of a repulsive lone pair interaction.
Sigma-Aldrich
D-Ala-D-Ala
Pricing and availability is not currently available.
Sigma-Aldrich
Ala-Ala
Pricing and availability is not currently available.