Merck
  • Metabolism of doxylamine succinate in Fischer 344 rats. Part III: Conjugated urinary and fecal metabolites.
Metabolism of doxylamine succinate in Fischer 344 rats. Part III: Conjugated urinary and fecal metabolites.
Journal of analytical toxicology (1990-07-11)
C L Holder, P H Siitonen, W Slikker, C J Branscomb, W A Korfmacher, H C Thompson, C E Cerniglia, A B Gosnell, J O Lay
ABSTRACT
Elimination and metabolic profiles of the glucuronide products of doxylamine and its N-demethylated metabolites were determined after the oral administration of (14C)-doxylamine succinate (13.3 and 133 mg/kg doses) to male and female Fischer 344 rats. The cumulative urinary and fecal eliminations of these conjugated doxylamine metabolites at the 13.3 mg/kg dose were 44.4 +/- 4.2% and 47.3 +/- 8.1% of the total recovered dose for male and female rats, respectively. The cumulative urinary and fecal eliminations of conjugated doxylamine metabolites at the 133 mg/kg dose were 55.2 +/- 2.6% and 47.9 +/- 2.5% of the total recovered dose for male and female rats, respectively. The conjugated doxylamine metabolites that were isolated, quantitated, and identified are doxylamine O-glucuronide, N-desmethyl-doxylamine O-glucuronide, and N,N-didesmethyldoxylamine O-glucuronide.
Sigma-Aldrich
Doxylamine succinate salt
Supelco
Doxylamine succinate solution, 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®