Merck
  • Development of enzyme-linked immunosorbent assays for Sudan dyes in chilli powder, ketchup and egg yolk.
Development of enzyme-linked immunosorbent assays for Sudan dyes in chilli powder, ketchup and egg yolk.
Food additives & contaminants. Part A, Chemistry, analysis, control, exposure & risk assessment (2009-08-15)
Laura Anfossi, Claudio Baggiani, Cristina Giovannoli, Gianfranco Giraudi
ABSTRACT
This study aimed at developing sensitive competitive enzyme-linked immunosorbent assays (ELISAs) for the banned Sudan dyes using polyclonal antibodies. Three different formats were developed and characterized in terms of sensitivity, selectivity and rapidity. A competitive indirect ELISA was developed, which showed an IC(50) of 3.8 microg l(-1). Two competitive direct ELISAs were also developed, in which the antibody was added before or simultaneously with the other reagents; the first showed an IC(50) of 8.3 microg l(-1) and the latter showed an IC(50) of 4.9 microg l(-1). Nevertheless, considering dilution of extracts which is needed to offset matrix interference, the limits of detection of the three formats were substantially the same (10 microg kg(-1)). The antibodies in all three test formats were able to recognize Sudan I and partially Sudan II, III and IV; no cross-reactivity was observed with the five edible dyes. Twenty food samples, including chilli powder, paprika, ketchup, and egg, were extracted by a simple sample preparation and very limited dilution. Extracts were analyzed by the developed competitive direct ELISA with the simultaneous addition of reagents. A good correlation was observed (y = 1.19 x-10.0, r(2) = 0.991, n = 20) when the data was compared with that obtained through a conventional HPLC method.
Sigma-Aldrich
Sudan III, BioXtra, certified by the Biological Stain Commission
Sigma-Aldrich
Sudan III, Technical grade
Supelco
Sudan III, analytical standard
Supelco
Sudan IV, analytical standard
Sigma-Aldrich
Sudan IV, Certified Biological Stain Commission,Dye content ≥80 %
Sigma-Aldrich
Sudan II, Dye content 90 %
Sigma-Aldrich
Sudan IV, BioXtra, certified by the Biological Stain Commission
Supelco
Sudan II, analytical standard