- Solvent-promoted and -controlled aza-Michael reaction with aromatic amines.
Solvent-promoted and -controlled aza-Michael reaction with aromatic amines.
The Journal of organic chemistry (2009-07-23)
Kavita De, Julien Legros, Benoit Crousse, Danièle Bonnet-Delpon
PMID19621888
ABSTRACT
1,4-Addition of anilines onto Michael acceptors proceeds easily in specific polar protic solvents, without any promoting agent. According to the solvent and to the electrophile, the selectivity of the reaction can be finely tuned. With methyl acrylate as electrophile, only monoaddition takes place in water, while the diadduct is yielded in hexafluoroisopropyl alcohol (HFIP). The use of methyl vinyl ketone as a partner affords the monoadduct in water, the diadduct in trifluoroethanol (TFE), and the quinoline in HFIP.
Sigma-Aldrich
3-Buten-2-one, contains 0.3-1.0% hydroquinone as stabilizer, technical grade, 90%