Merck
  • Effects of polychlorinated biphenyls on the neutrophil NADPH oxidase system.
Effects of polychlorinated biphenyls on the neutrophil NADPH oxidase system.
Toxicology letters (2009-05-12)
Oddvar Myhre, Espen Mariussen, Trine Reistad, Oyvind Albert Voie, Halvor Aarnes, Frode Fonnum
ABSTRACT
Polychlorinated biphenyls (PCBs) are reported to induce the formation of reactive oxygen species (ROS) in human neutrophil granulocytes through the activation of the NADPH oxidase. The purpose of the present study is to elucidate the cellular mechanisms responsible for the activation of the NADPH oxidase after exposure to PCB. We have previously shown that PCB activates human neutrophil granulocytes through a calcium dependent activation of phospholipase D and/or phospholipase C, followed by the activation of protein kinase C. In the present study, pharmacological characterization of Aroclor (A) 1242-induced respiratory burst in human neutrophils was conducted by the use of enzymatic inhibitors. Pre-incubation with U0126, SB203580, SP600125, cyclosporin A and FK506 attenuated the A 1242-induced respiratory burst, measured by DCF-fluorescence, and luminol-amplified chemiluminescence. Our results show that the Erk1/2 kinases and p38MAPK/JNK are involved in ROS formation in neutrophils exposed to A 1242.
Product Description
Supelco
Aroclor 1242 solution, certified reference material, 500 mg/kg in transformer oil
Supelco
Aroclor 1242 solution, certified reference material, 50 mg/kg in transformer oil