- Poly(lactide-co-glycolide) microspheres: a potent oral delivery system to elicit systemic immune response against inactivated rabies virus.
Poly(lactide-co-glycolide) microspheres: a potent oral delivery system to elicit systemic immune response against inactivated rabies virus.
Rabies is an endemic, fatal zoonotic disease in the developing countries. Oral vaccination strategies are suitable for rabies control in developing countries. Studies were performed to investigate the suitability of poly(lactide-co-glycolide) (PLG) microspheres as an oral delivery system for beta-propiolactone inactivated concentrated rabies virus (CRV). Immune responses induced by encapsulated (PLG+CRV) and un-encapsulated inactivated rabies virus after oral and intraperitoneal route administrations were compared. The anti-rabies virus IgG antibody titer, virus neutralizing antibody (VNA) titers obtained by mouse neutralization test (MNT) and IgG2a and IgG1 titers of mice group immunized orally with PLG+CRV showed significantly (p<0.001) higher response than the group immunized orally with un-encapsulated CRV. There was no significant difference (p>0.05) between groups inoculated by intraperitoneal route. The stimulation index (SI) obtained by lymphoproliferation assay of PLG+CRV oral group also showed significantly (p<0.001) higher response than the group immunized orally with un-encapsulated CRV, suggesting that oral immunization activates Th1-mediated cellular immunity. Immunized mice of all experimental groups were challenged intracerebrally with a lethal dose of virulent rabies virus Challenge Virus Standard (CVS). The survival rates of mice immunized orally with PLG+CRV and CRV alone were 75% and 50%, respectively, whereas intraperitoneally immunized groups showed 100% protection. The overall results of humoral, cellular immune response and survival rates of mice immunized orally with PLG+CRV were significantly (p<0.001) higher than those of mice immunized orally with CRV alone. These data suggest that the PLG encapsulated inactivated rabies virus can be used for oral immunization against rabies.