Merck
  • Highly efficient copper-catalyzed domino ring opening and Goldberg coupling cyclization for the synthesis of 3,4-dihydro-2H-1,4-benzoxazines.
Highly efficient copper-catalyzed domino ring opening and Goldberg coupling cyclization for the synthesis of 3,4-dihydro-2H-1,4-benzoxazines.
Organic letters (2009-04-04)
R Koteshwar Rao, Ajay B Naidu, Govindasamy Sekar
ABSTRACT
trans-3,4-Dihydro-2H-1,4-benzoxazine moieties can be synthesized by domino aziridine ring opening with o-iodophenols followed by the copper-catalyzed Goldberg coupling cyclization (intramolecular C(aryl)-N(amide) bond formation) with good to excellent yields.
Sigma-Aldrich
2-Iodophenol, 98%