Merck
  • Preparation of achiral and chiral (E)-enaminopyran-2,4-diones and their phytotoxic activity.
Preparation of achiral and chiral (E)-enaminopyran-2,4-diones and their phytotoxic activity.
Journal of agricultural and food chemistry (2009-01-28)
Luiz C Dias, Antônio J Demuner, Vânia M M Valente, Luiz C A Barbosa, Felipe T Martins, Antônio C Doriguetto, Javier Ellena
ABSTRACT
A short and efficient approach to a range of new chiral and achiral functionalized (E)-enaminopyran-2,4-diones starting with commercially available dehydroacetic acid is described. The phytotoxic properties of these (E)-enaminopyran-2,4-diones were evaluated by their ability to interfere with the growth of Sorghum bicolor and Cucumis sativus seedlings. A different sensitivity of the two crops was evident with the (E)-enaminopyran-2,4-diones. The most active compounds were also tested against two weeds, Ipomoea grandifolia and Brachiaria decumbens. To the best of our knowledge, this is the first report describing enaminopyran-2,4-diones as potential plant growth regulators.
Sigma-Aldrich
Dehydroacetic acid, ≥98.0% (T)