Merck
  • Self-assembled monolayers of 2-(thienyl)hexylphosphonic acid on native oxide surface of silicon fabricated by air-liquid interface-assisted method.
Self-assembled monolayers of 2-(thienyl)hexylphosphonic acid on native oxide surface of silicon fabricated by air-liquid interface-assisted method.
Journal of colloid and interface science (2008-06-03)
Chih-Wei Hsu, Huei-Ru Liou, Wei-Fang Su, Leeyih Wang
ABSTRACT
A simple, fast, and low-compound-consuming procedure based on the air-liquid interface-assisted method for preparing self-assembled monolayers (SAMs) of organic molecules with phosphonic acid head groups on the native oxide surface of silicon was demonstrated. The SAMs thus prepared were characterized by contact angle measurement, X-ray photoelectron spectroscopy (XPS), Fourier transform infrared spectroscopy (FTIR), and atomic force microscopy (AFM). This approach enabled the fabrication of ordered SAMs in a large-area substrate.
Sigma-Aldrich
Silicon monoxide, powder, −325 mesh
Sigma-Aldrich
Silicon monoxide, pieces, 3-10 mm, 99.99% trace metals basis