MilliporeSigma
  • Hydroquinone dioxygenase from pseudomonas fluorescens ACB: a novel member of the family of nonheme-iron(II)-dependent dioxygenases.
Hydroquinone dioxygenase from pseudomonas fluorescens ACB: a novel member of the family of nonheme-iron(II)-dependent dioxygenases.
Journal of bacteriology (2008-05-27)
Mariëlle J H Moonen, Silvia A Synowsky, Willy A M van den Berg, Adrie H Westphal, Albert J R Heck, Robert H H van den Heuvel, Marco W Fraaije, Willem J H van Berkel
ABSTRACT
Hydroquinone 1,2-dioxygenase (HQDO), an enzyme involved in the catabolism of 4-hydroxyacetophenone in Pseudomonas fluorescens ACB, was purified to apparent homogeneity. Ligandation with 4-hydroxybenzoate prevented the enzyme from irreversible inactivation. HQDO was activated by iron(II) ions and catalyzed the ring fission of a wide range of hydroquinones to the corresponding 4-hydroxymuconic semialdehydes. HQDO was inactivated by 2,2'-dipyridyl, o-phenanthroline, and hydrogen peroxide and inhibited by phenolic compounds. The inhibition with 4-hydroxybenzoate (K(i) = 14 microM) was competitive with hydroquinone. Online size-exclusion chromatography-mass spectrometry revealed that HQDO is an alpha2beta2 heterotetramer of 112.4 kDa, which is composed of an alpha-subunit of 17.8 kDa and a beta-subunit of 38.3 kDa. Each beta-subunit binds one molecule of 4-hydroxybenzoate and one iron(II) ion. N-terminal sequencing and peptide mapping and sequencing based on matrix-assisted laser desorption ionization--two-stage time of flight analysis established that the HQDO subunits are encoded by neighboring open reading frames (hapC and hapD) of a gene cluster, implicated to be involved in 4-hydroxyacetophenone degradation. HQDO is a novel member of the family of nonheme-iron(II)-dependent dioxygenases. The enzyme shows insignificant sequence identity with known dioxygenases.
Sigma-Aldrich
4′-Hydroxyacetophenone, 99%