Merck
  • Bio-inspired polyene cyclization: synthesis of tetracyclic terpenoids promoted by steroidal acetal-SnCl4.
Bio-inspired polyene cyclization: synthesis of tetracyclic terpenoids promoted by steroidal acetal-SnCl4.
Chemical communications (Cambridge, England) (2008-03-14)
Yu-Jun Zhao, Teck-Peng Loh
ABSTRACT
This communication describes a highly efficient intermolecular polyene cyclization method using steroidal acetals as the initiators to synthesize tetracyclic terpenoids; both good yields and good asymmetric induction were obtained.
Sigma-Aldrich
Tin(IV) chloride, 99.995% trace metals basis
Sigma-Aldrich
Tin(IV) chloride solution, 1.0 M in methylene chloride
Sigma-Aldrich
Tin(IV) chloride, 98%