Merck
  • [Studies on flavonoid constituents from herbs of Artemisia ordosica II].
[Studies on flavonoid constituents from herbs of Artemisia ordosica II].
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica (2007-03-14)
Wei Zhang, Dong-bao Zhao, Ming-jing Li, Xiu-hua Liu, Han-qing Wang
ABSTRACT
To study the chemical constituents of herbs of Artemisia ordosica. The chemical constituents were isolated and repeatedly purified on silica gel column and the structures were elucidated by the NMR spectra and physicochemical properties. Eight flavones were obtained and they were identified as isosakuranetin, 7, 4'-dimethylaro madendrin, acacetin, cirsimaritin, rhamnetin, eupatolin, 5, 7, 2', 4'-tetrahydroxy-6, 5'-dimethoxyflavone, hyperoside. All the compounds were obtained from herbs of A. ordosica for the first time.
Product Description
Supelco
Rhamnetin, analytical standard
Sigma-Aldrich
Cirsimaritin, ≥90% (LC/MS-ELSD)