Merck
  • Postirradiation electron transfer vs differential radical decay in X-irradiated DNA and its mixtures with additives. Electron spin resonance spectroscopy in LiBr glass at low temperatures.
Postirradiation electron transfer vs differential radical decay in X-irradiated DNA and its mixtures with additives. Electron spin resonance spectroscopy in LiBr glass at low temperatures.
The journal of physical chemistry. B (2006-07-28)
Chandrima Pal, Jürgen Hüttermann
ABSTRACT
Free radical formation in DNA and in colyophilized mixtures of DNA with the additives mitoxantrone and riboflavin was monitored after X-ray irradiation in frozen aqueous glasses (7 M LiBr/D2O) at 77 K by electron spin resonance (ESR) spectroscopy. Specifically, the postirradiation time course at 77 K of the respective free radical intensity residing on DNA or on the additive was probed in order to test the hypothesis of electron transfer from DNA, e.g., to mitoxantrone after irradiation under these conditions (e.g., Messer, A.; Carpenter, K.; Forzley, K.; Buchanan, J.; Yang, S.; Razskazovskii, Y.; Cai, Z.; Sevilla, M. D. J. Phys. Chem. B 2000, 104, 1128). For both additives, different additive loadings and irradiation doses were employed. The observed relative change in contributions of DNA and of additive radical components to the experimental spectra with time could be ascribed, for both additives, unequivocally to independent, differential fading of component radicals. Transfer from DNA to the additive, e.g., by electron tunneling as proposed before could be ruled out to occur by a detailed, quantitative analysis of the experimental spectra using reconstruction techniques. Additional studies were performed with the nucleotides TMP and dCMP and its mixtures with mitoxantrone in order to describe the time course in systems which are expected to behave independently; the results supported the conclusions arrived at from the analysis of the DNA/additive system. A model was proposed to describe the postirradiation radical fading mechanisms which involve liberation of radiation-induced matrix-trapped defects with time. It was assumed that these defects are ESR-mute and react with radicals by net radical destruction. Some experimental observations are presented concerning influence of temperature and of the matrix on the fading processes. These seem to argue in favor of such a model although a detailed, quantitative description is still not possible.
Sigma-Aldrich
Lithium bromide solution, 54 wt. % in H2O
Sigma-Aldrich
Lithium bromide, ReagentPlus®, ≥99%
Sigma-Aldrich
Lithium bromide, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Lithium bromide, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis
Sigma-Aldrich
Lithium bromide, powder and chunks, ≥99.995% trace metals basis